Anti-KCC1/SLC12A4 Goat Polyclonal Antibody
89296-962
: Genetex
- Antibody Type:Primary
- Antigen Name:Solute Carrier Family 12 (Potassium/Chloride Transporter), Member 4
- Antigen Symbol:KCC1/SLC12A4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:6560
- Antigen Synonyms:KCC1
- Storage Buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage Temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5mg/ml
- Immunogen:C-DPSHAPDNFRE
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:89296-962
- Supplier no.:GTX88934
Goat polyclonal antibody to KCC1 / SLC12A4
Approx 150kDa band observed in Human Heart and Skeletal Muscle lysates (calculated MW of 121kDa according to NP_005063.1). Recommended concentration: 0.3-1µg/ml.
Type: Primary
Antigen: KCC1/SLC12A4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Dog, Rat