Anti-AF4 Rabbit Polyclonal Antibody
89290-496
: Genetex
- Antibody Type:Primary
- Antigen Name:AF4/FMR2 Family, member 1
- Antigen Symbol:AF4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Size:100 μg
- Gene ID:4299
- Storage Buffer:Phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Storage Temperature:Store at -20 degrees Celcius. Product is stable for at least 1 year.
- Immunogen:The antiserum was produced against synthesized peptide derived from human AF4.
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen.
- Cat. No.:89290-496
- Supplier no.:GTX87470
Rabbit polyclonal antibody to AF4
We recommend the following starting dilutions: IHC: 1:50~1:100 ELISA: 1:20000Optimal dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: AF4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse