Anti-FOXD4/L2/L3/L4/L5/L6 Rabbit Polyclonal Antibody
Catalog # 89290-038
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Forkhead box D4-like 6
- Antigen Symbol:FOXD4/L2/L3/L4/L5/L6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:653404
- Storage Buffer:Phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Storage Temperature:Store at -20 degrees Celcius. Product is stable for at least 1 year.
- Immunogen:The antiserum was produced against synthesized peptide derived from human FOXD4/L2/L3/L4/L5/L6.
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen.
- Cat. No.:89290-038
- Supplier no.:GTX87238
Specifications
About this item
Rabbit polyclonal antibody to FOXD4/L2/L3/L4/L5/L6
We recommend the following starting dilutions: WB: 1:500~1:1000 ELISA: 1:40000Optimal dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: FOXD4L6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human