To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Protein Kinase D2
- Antigen Symbol:PRKD2
- Clonality:Monoclonal
- Clone:3B12
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Gel Shift:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Monkey,Human,Dog,Rat
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:25865
- Antigen Synonyms:PRKCM|HSPC187|Protein kinase D|PKCu|Serine/threonine-protein kinase D2|nPKC-D2|nPKC-D1|Protein kinase C nu type|PKC nu|PRKCN|PKC-u|PKC-nu|PKC mu|PKD|nPKC-nu|PKC-v|PKD3|Serine/threonine-protein kinase D3|PRKD2|Serine/threonine-protein kinase D1|PKCv|PKD1|Protein kinase C mu type|EPK2|PKCmu|PKC v|PKC u|PRKD3|Protein kinase EPK2|PRKD1|PKD2|PKCnu|nPKC-mu
- Storage Buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0.02% sodium azide.
- Storage Temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human PRKD2 (NP_057541) produced in HEK293T cell.
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Cat. No.:89272-440
- Supplier no.:GTX83819
Mouse monoclonal antibody [3B12] to PRKD2
Recommended Starting Dilutions: WB 1:2000 ICC/IF 1:100 Flow Cytometry 1:100Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: PRKD2
Clonality: Monoclonal
Clone: 3B12
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Dog, Monkey, Rat