To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Myocyte Enhancer Factor 2C
- Antigen Symbol:MEF2C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:400 μl
- Gene ID:4208
- Antigen Synonyms:MADS box transcription enhancer factor 2 polypeptide C myocyte enhancer factor 2C|Myocyte enhancer factor 2C|Similar to MADS box transcription enhancer factor 2 polypeptide C|MEF2Cphospho S387|MEF2C_HUMAN.|Myocyte specic enhancer factor 2C
- Storage Buffer:PBS with 0.09% (W/V) sodium azide
- Storage Temperature:Store at at 2-8°C for up to 6 months. For long term storage, aliquot and store at -20 °C. Avoid repeated freeze/thaw cycles.
- Concentration:0.25mg/ml
- Immunogen:KLH conjugated synthetic peptide corresponding to amino acid residues surrounding T300 of human MEF2C.
- Purification:Purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:89302-908
- Supplier no.:GTX81913
Specifications
About this item
Rabbit polyclonal to MEF2C (T300)
Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:50-100IHC: Use at 1:10-50Optimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: MEF2C
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human