To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Sperm Associated Antigen 9
- Antigen Symbol:JLP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:400 μl
- Environmentally Preferable:
- Gene ID:9043
- Antigen Synonyms:PIG6|PHET|HLC6|HLC-6|JLP|CT89|JIP4|JIP-4|HLC4
- Storage Buffer:PBS with 0.09% (W/V) sodium azide
- Storage Temperature:Store at at 2-8°C for up to 6 months. For long term storage, aliquot and store at -20 °C. Avoid repeated freeze/thaw cycles.
- Immunogen:The immunogen corresponding to a region within amino acids 603 and 632 of human SPAG9.
- Purification:Prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis against PBS.
- Cat. No.:89302-180
- Supplier no.:GTX81547
Specifications
About this item
Rabbit polyclonal to JLP (SPAG9) (Internal)
Recommended Starting Dilutions: ForWB: Use at a dilution of 1:1000. For IHC-P: Use at a dilution of 1:10-50. Optimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: JLP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human