Anti-SLC22A1 Mouse Monoclonal Antibody [clone: 2C5]
Catalog # 89291-678
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Solute Carrier Family 22 Member A1
- Antigen Symbol:SLC22A1
- Clonality:Monoclonal
- Clone:2C5
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Gel Shift:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:6580
- Antigen Synonyms:HOCT1|SLC22A1|OCT1|oct1_cds
- Storage Buffer:Ascitic fluid containing 0.03% sodium azide.
- Storage Temperature:Store at 4℃, for long term storage, store at -20℃.
- Immunogen:The immunogen corresponding to human SLC22A1 expressed in E. Coli.
- Cat. No.:89291-678
- Supplier no.:GTX80400
Specifications
About this item
Mouse Monoclonal antibody [2C5] to SLC22A1
Recommended Starting Dilutions:ELISA: 1/10000.WB: 1/500 - 1/2000.FCM: 1/200 - 1/400.Not yet tested in other applications.Determining optimal working dilutions by titration test.
Type: Primary
Antigen: SLC22A1
Clonality: Monoclonal
Clone: 2C5
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human