Anti-SIT1 Mouse Monoclonal Antibody [clone: SIT-01]
Catalog # 89284-656
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Signaling Threshold Regulating Transmembrane Adaptor 1
- Antigen Symbol:SIT1
- Clonality:Monoclonal
- Clone:SIT-01
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:27240
- Antigen Synonyms:SIT|SIT1_HUMAN|SHP intracting transmembrane adapter protein|Signaling threshold-regulating transmembrane adapter 1|gp30/40|SHP2-interacting transmembrane adapter protein|Sit1|Suppression-inducing transmembrane adapter 1.|Suppression inducing transmembrane adapter 1
- Storage Buffer:The reagent is provided in phosphate buffered saline (PBS) containing 15 mM sodium azide and 0.2% (w/v) high-grade protease free Bovine Serum Albumin (BSA) as a stabilizing agent.
- Storage Temperature:Store in the dark at 2-8°C. Do not freeze. Do not use after expiration date stamped on vial label. Short-term exposure to room temperature should not affect the quality of the reagent.
- Immunogen:Bacterially produced recombinant intracellular fragment of human SIT.
- Cat. No.:89284-656
- Supplier no.:GTX79892
Specifications
About this item
PE-conjugated Mouse Monoclonal antibody [SIT-01] to SIT
Recommended Starting Dilutions:FACS: Staining technique:intracellular staining Application noteWB: Note: Western blot analysis demonstrated that SIT is expressed as an approximately 40 kDa protein that is reduced to approximately 20 kDa by endoglycosidase treatment.Optimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: SIT1
Clonality: Monoclonal
Clone: SIT-01
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human