Anti-SOS2 Mouse Monoclonal Antibody [clone: SOS-01]
89299-984
: Genetex
- Antibody Type:Primary
- Antigen Name:Son Of Sevenless Homolog 2
- Antigen Symbol:SOS2
- Clonality:Monoclonal
- Clone:SOS-01
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:6655
- Storage Buffer:Phosphate buffered saline (PBS) with 15 mM sodium azide, approx. pH 7.4
- Storage Temperature:Store at 2-8°C. Do not use after expiration date stamped on vial label. For long-term storage aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Immunogen:Peptide corresponding to amino acids THPSMHRDGPPLLENAHSS of human Sos protein.
- Cat. No.:89299-984
- Supplier no.:GTX79891
Mouse Monoclonal antibody [SOS-01] to Sos
Recommended Starting Dilutions:WB: Use at 1 µg/mL. Positive Control: HeLa human cervix carcinoma cell line Use under reducing conditionsOptimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: SOS2
Clonality: Monoclonal
Clone: SOS-01
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse