Anti-CD62L Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: LT-TD180]
89285-602
: Genetex
- Antibody Type:Primary
- Antigen Name:L-Selectin
- Antigen Symbol:CD62L
- Clonality:Monoclonal
- Clone:LT-TD180
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 test
- Gene ID:6402
- Antigen Synonyms:LECAM1|SELL|CD62L|TQ1|LEU8|LNHR|LYAM1vPLNHR|LSEL|LAM1
- Storage Buffer:The reagent is provided in phosphate buffered saline (PBS) containing 15 mM sodium azide and 0.2% (w/v) high-grade protease free Bovine Serum Albumin (BSA) as a stabilizing agent.
- Storage Temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze. Keep protected from prolonged exposure to light.
- Immunogen:Peripheral blood leukocytes.
- Cat. No.:89285-602
- Supplier no.:GTX78400
Monoclonal Antibody to CD62L Fluorescein (FITC)
The reagent is designed for Flow Cytometry analysis of human blood cells using 20 ul reagent / 100 ul of whole blood or 106 cells in a suspension.The content of a vial (2 ml) is sufficient for 100 tests.
Type: Primary
Antigen: CD62L
Clonality: Monoclonal
Clone: LT-TD180
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: