Anti-CD43 (sialophorin) Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MEM-59]
Catalog # 89285-574
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Sialophorin
- Antigen Symbol:CD43 (sialophorin)
- Clonality:Monoclonal
- Clone:MEM-59
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 test
- Gene ID:6693
- Antigen Synonyms:LSN|CD43|GPL115|GALGP
- Storage Buffer:The reagent is provided in phosphate buffered saline (PBS) containing 15 mM sodium azide and 0.2% (w/v) high-grade protease free Bovine Serum Albumin (BSA) as a stabilizing agent.
- Storage Temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze. Keep protected from prolonged exposure to light.
- Immunogen:Human T lymphocytes.
- Cat. No.:89285-574
- Supplier no.:GTX78289
Specifications
About this item
Monoclonal Antibody to CD43 (Human) Fluorescein (FITC)
The reagent is designed for Flow Cytometry analysis of human blood cells using 20 ul reagent / 100 ul of whole blood or 106 cells in a suspension.The content of a vial (2 ml) is sufficient for 100 tests.
Type: Primary
Antigen: CD43 (sialophorin)
Clonality: Monoclonal
Clone: MEM-59
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: