Order Entry
United States
ContactUsLinkComponent
Anti-GPCR GPR85 (G Protein-coupled Receptor 85) Rabbit Polyclonal Antibody
Anti-GPCR GPR85 (G Protein-coupled Receptor 85) Rabbit Polyclonal Antibody
Catalog # 89278-414
Supplier:  Genetex
Anti-GPCR GPR85 (G Protein-coupled Receptor 85) Rabbit Polyclonal Antibody
Catalog # 89278-414
Supplier:  Genetex
Supplier Number:  GTX78037

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    G Protein-Coupled Receptor 85
  • Antigen Symbol:
    GPR85
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    25 μg
  • Gene ID:
    54329
  • Antigen Synonyms:
    SREB|SREB2
  • Storage Buffer:
    Phosphate buffered saline, pH 7.7, containing 0.01% sodium azide
  • Storage Temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Synthetic peptide - KLH conjugated. Peptide sequence identity/similarity to Mouse: 100/100%. Rat: 100/100%.
  • Purification:
    Peptide immunogen affinity column
  • Cat. No.:
    89278-414
  • Supplier no.:
    GTX78037

Specifications

About this item

Rabbit polyclonal to G Protein-Coupled Receptor GPR85/SREB2

Type: Primary
Antigen: GPR85
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human