Anti-GPR22 Rabbit Polyclonal Antibody
Catalog # 89278-186
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:G Protein-coupled Receptor 22
- Antigen Symbol:GPR22
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:25 μg
- Gene ID:2845
- Antigen Synonyms:Probable G-protein coupled receptor 22|G protein coupled receptor 22|GPCR GPR22.|GPR22|GPR22_HUMAN
- Storage Buffer:Phosphate buffered saline, PH 7.7 containing 0.01% sodium azide
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide - KLH conjugated
- Purification:Immunogen affinity purified.
- Cat. No.:89278-186
- Supplier no.:GTX71727
Specifications
About this item
Rabbit Polyclonal to Human G Protein-Coupled Receptor GPR22
Immunohistochemistry with formalin fixed paraffin embedded tissues. Not tested for other uses. IHC-P: Use at a dilution of 1/100 - 1/200.
Type: Primary
Antigen: GPR22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human