To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Cadherin, EGF Lag Seven-Pass G-Type Receptor 1
- Antigen Symbol:CELSR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:25 μg
- Gene ID:9620
- Antigen Synonyms:ME2|ADGRC1|CDHF9|HFMI2|FMI2
- Storage Buffer:Phosphate buffered saline, PH 7.7 containing 0.01% sodium azide
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide - KLH conjugated
- Purification:Immunogen affinity purified.
- Cat. No.:89278-146
- Supplier no.:GTX71611
Specifications
About this item
Rabbit Polyclonal to Human CELSR1
Immunohistochemistry with formalin fixed paraffin embedded tissues. Not tested for other uses. IHC-P: Use at a dilution of 1/100 - 1/200.
Type: Primary
Antigen: CELSR1 (Cadherin EGF LAG seven-pass G-typeR 1 (flamingo homolog Drosophila))
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: