To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Sodium/Calcium Exchanger
- Antigen Symbol:SLC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:65956
- Antigen Synonyms:Secondary lymphoid-tissue chemokine|Exodus 2|C-C motif chemokine 21|TCA4|Exodus-2|Exodus2|Small-inducible cytokine A21|6Ckine|NKX|Na+/C2+ exchanger
- Storage Buffer:PBS (pH 7.2) containing 50% glycerol, 1% BSA, 0.02% sodium azide
- Storage Temperature:The antibody solution should be gently mixed before use. Stored at -20 °C for future use.
- Concentration:0.5 mg/ml
- Purification:Antigen affinity purification
- Cat. No.:89283-644
- Supplier no.:GTX59875
Specifications
About this item
Rabbit Polyclonal antibody to CCL21
Western blot analysis (0.5-1 µg/ml). However, the optimal conditions should be determined individually. Cross-reactivity to SLC of other species has not been determined.
Type: Primary
Antigen: SLC (Chemokine (C-C motif) ligand 21C (leucine))
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat