To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Calcium/Calmodulin-dependent Protein Kinase II alpha
- Antigen Symbol:CAMK2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 μl
- Environmentally Preferable:
- Gene ID:815
- Antigen Synonyms:calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha|calcium/calmodulin-dependent protein kinase II alpha-B subunit|calcium/calmodulin-dependent protein kinase II alpha|Calci|calcium/calmodulin-dependent protein kinase type II alpha chain
- Modification:phospho-Thr286
- Storage Buffer:PBS (pH 7.2) containing 50% glycerol, 1% BSA, and 0.02% thimerosal
- Storage Temperature:The antibody solution should be gently mixed before use. Store at +4C. Product should remain stable for at least 12 months.
- Purification:Antigen affinity purification
- Cat. No.:89299-942
- Supplier no.:GTX59564
Specifications
About this item
Rabbit polyclonal antibody to CaMKII (phospho Thr286)
Western blotting at 1:500-1:1000. However, the optimal concentrations should be determined individually. The anti-Phospho-CaMKII antibody recognizes only the phosphorylated CaMKII (Thr286) in samples from human, mouse, and rat origins.
Type: Primary
Antigen: CaMKII
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat