Anti-Pbk1 (ribosomal L1 Domain containing 1) Rabbit Polyclonal Antibody
Catalog # 89317-552
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Ribosomal L1 Domain Containing 1
- Antigen Symbol:Pbk1 (ribosomal L1 Domain containing 1)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 μg
- Gene ID:26156
- Antigen Synonyms:UTP30|CSIG|L12|PBK1
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 and 0.01% (w/v) Sodium Azide
- Storage Temperature:Store vial at –20 °C prior to opening. Aliquot contents and freeze at –20 °C or below for extended storage Avoid cycles of freezing and thawing.
- Concentration:1.1 mg/ml
- Immunogen:This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal sequence of human PBK1.
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89317-552
- Supplier no.:GTX48683
Specifications
About this item
Rabbit Polyclonal antibody to Pbk1
Recommended Starting Dilutions: WB: Use at 1:500 - 1:2,000 ELISA: Use at 1:2,500 - 1:10,000 IP: Use at 1:100
Type: Primary
Antigen: Pbk1 (ribosomal L1 Domain containing 1)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human