Anti-CDH22 (Cadherin 22 type 2) Rabbit Polyclonal Antibody
Catalog # 89288-424
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Cadherin 22
- Antigen Symbol:CDH22
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:64405
- Antigen Synonyms:C20orf25|dJ998H6.1
- Storage Buffer:Phosphate buffered saline (PBS)
- Storage Temperature:Store at 4 °C for short-term use. For longer periods of storage, aliquot and store at –20 °C. Avoid repeat freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide
- Purification:Protein A Purified Total IgG
- Cat. No.:89288-424
- Supplier no.:GTX46027
Specifications
About this item
Rabbit polyclonal antibody to CDH22 (N-terminal)
Recommended Dilutions: For Western Blot: Use at 5.0 ug/ml.
Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: CDH22 (Cadherin 22 type 2)
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: