Anti-CCL21 (chemokine (C-C motif) ligand 21b) Rabbit Polyclonal Antibody
89337-268
: Genetex
- Antibody Type:Primary
- Antigen Name:CCL21
- Antigen Symbol:CCL21 (chemokine (C-C motif) ligand 21b)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat
- Western Blot:Yes
- Size:100 μg
- Gene ID:298006
- Storage Buffer:Phosphate buffered saline, pH 7.2, containing 0.1% sodium azide
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant protein - rat CCL21
- Purification:Protein A affinity purified
- Cat. No.:89337-268
- Supplier no.:GTX45119
Rabbit polyclonal to CCL21
For WB: Use at a dilution of 1:5,000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: CCL21 (chemokine (C-C motif) ligand 21b)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat