To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG
- Clonality:Monoclonal
- Clone:MK 1 A6
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Macaque,Human
- Size:100 μg
- Environmentally Preferable:
- Epitope:(Fc) CH2 Domain
- Gene ID:3500
- Storage Buffer:Phosphate buffered saline, 0.01% Thiomersal
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Human IgG Polyclonal
- Cat. No.:89283-140
- Supplier no.:GTX43308
Specifications
About this item
HRP-conjugated Mouse Monoclonal antibody [MK 1 A6] to Human IgG (Fc) CH2 Domain
Recommended Starting Dilutions:
Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: MK 1 A6
Conjugation:
Epitope: (Fc) CH2 Domain
Host: Mouse
Isotype:
Reactivity: Human, Rhesus Monkey