Order Entry
United States
ContactUsLinkComponent
Anti-CD34 Mouse Monoclonal Antibody [clone: 1H6]
Anti-CD34 Mouse Monoclonal Antibody [clone: 1H6]
Catalog # 89265-426
Supplier:  Genetex
Anti-CD34 Mouse Monoclonal Antibody [clone: 1H6]
Catalog # 89265-426
Supplier:  Genetex
Supplier Number:  GTX43103
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    CD34 molecule
  • Antigen Symbol:
    CD34
  • Clonality:
    Monoclonal
  • Clone:
    1H6
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Dog
  • Size:
    100 Tests
  • Environmentally Preferable:
  • Antigen Synonyms:
    CD34 antigen|Mucosialin|hematopoietic progenitor cell antigen CD34|RP11-328D5.2
  • Cat. No.:
    89265-426
  • Supplier no.:
    GTX43103

Specifications

About this item

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 1H6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: