To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Stathmin 1
- Antigen Symbol:STMN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 μl
- Environmentally Preferable:
- Gene ID:3925
- Antigen Synonyms:Op18|SMN|C1orf215|PP19|LAP18|OP18|Stathmin|Lag|PR22|Leukemia-associated phosphoprotein p18|PP17
- Modification:phospho-Ser38
- Storage Buffer:In phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Storage Temperature:Aliquot and store at –20 °C or –80 °C. Avoid repeated freeze/thaw cycles.
- Concentration:1 mg/ml
- Immunogen:The antiserum was produced against synthesized phosphopeptide derived from human Stathmin 1 around the phosphorylation site of serine 38 (P -L-SP-P-P).
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific phosphopeptide. The antibody against non-phosphopeptide was removed by chromatography using non-phosphopeptide corresponding to the phosphorylation
- Cat. No.:89282-028
- Supplier no.:GTX38638
Specifications
About this item
Rabbit polyclonal antibody to Stathmin 1 (phospho Ser38)
WB: 1:500-1:1000 IHC: 1:50-1:100
Type: Primary
Antigen: STMN1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat