Anti-MOG Mouse Monoclonal Antibody
Catalog # 89282-302
Supplier: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Myelin Oligodendrocyte Glycoprotein
- Antigen Symbol:MOG
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:4340
- Antigen Synonyms:BTNL11|NRCLP7|MOGIG2|BTN6
- Storage Buffer:PBS pH 7.3. Contains 0.09 % sodium azide
- Storage Temperature:Store at 4oC for short term and at -20oC for long term storage.
- Immunogen:synthetic pMOG 35-55.
- Purification:Ion exchange column.
- Cat. No.:89282-302
- Supplier no.:GTX37234
Specifications
About this item
Mouse Monoclonal antibody to MOG
Type: Primary
Antigen: MOG
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human