Order Entry
United States
ContactUsLinkComponent
Anti-PI4K2B Rabbit Polyclonal Antibody
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    PI 4 Kinase 2 beta (phosphatidylinositol 4-kinase type 2 beta)
  • Antigen Symbol:
    PI4K2B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    400 μl
  • Environmentally Preferable:
  • Gene ID:
    55300
  • Antigen Synonyms:
    PIK42B|PI4KIIB
  • Storage Buffer:
    Preservative: 0.09% Sodium Azide; Constituents: PBS
  • Storage Temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    0.25 mg/ml
  • Immunogen:
    Synthetic peptide conjugated to KLH, corresponding to amino acids 319-336 of Human PI4K II beta .
  • Purification:
    Protein G purified
  • Cat. No.:
    89279-050
  • Supplier no.:
    GTX25462

Specifications

About this item

Rabbit polyclonal to PI 4 Kinase type 2 beta

ELISA: Use at a dilution of 1/1000. WB: Use at a dilution of 1/100 - 1/500. Detects a band of approximately 55 kDa (predicted molecular weight: 57 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PI4K2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse