Anti-GPR25 Rabbit Polyclonal Antibody
Catalog # 89277-636
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:G Protein-coupled Receptor 25
- Antigen Symbol:GPR25
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:25μg
- Gene ID:2848
- Antigen Synonyms:Probable G protein coupled receptor GPR25|G protein coupled receptor 25|GPR25_HUMAN|Probable G-protein coupled receptor 25|Gpr25|GPCR GPR25.
- Storage Buffer:Preservative: 0.01% Sodium Azide; Constituents: PBS. pH 7.7
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide (unfortunately, the amino acid sequence is considered to be commercially sensitive)
- Purification:Immunogen affinity purified
- Cat. No.:89277-636
- Supplier no.:GTX12691
Specifications
About this item
IHC-P: Use at a dilution of 1/100 - 1/200. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: GPR25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human