Order Entry
United States
ContactUsLinkComponent
Anti-EX33 Rabbit Polyclonal Antibody
Anti-EX33 Rabbit Polyclonal Antibody
Catalog # 89277-280
Supplier:  Avantor
Anti-EX33 Rabbit Polyclonal Antibody
Catalog # 89277-280
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Gene ID:
    53831
  • Storage Buffer:
    Preservative: 0.01% Sodium Azide; Constituents: PBS. pH 7.7
  • Storage Temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Synthetic peptide (unfortunately, the amino acid sequence is considered to be commercially sensitive)
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    89277-280
  • Supplier no.:
    GTX12681

Specifications

About this item

IHC-P: Use at a dilution of 1/100 - 1/200. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: GPR84
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human