To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Synovial Apoptosis Inhibitor 1, Synoviolin
- Antigen Symbol:HRD1(SYNV1) (synovial apoptosis inhibitor 1, synoviolin)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:100 μl
- Gene ID:84447
- Antigen Synonyms:HRD1|DER3
- Storage Buffer:1XPBS, 1% BSA, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human HRD1(SYNV1). The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89313-874
- Supplier no.:GTX118990
Specifications
About this item
Antibody reactive against peptide, detects immunogen. Further validation in progress.
Type: Primary
Antigen: HRD1(SYNV1) (synovial apoptosis inhibitor 1, synoviolin)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: