Order Entry
United States
ContactUsLinkComponent
Anti-FAAP24 Rabbit Polyclonal Antibody
Anti-FAAP24 Rabbit Polyclonal Antibody
  89322-950
 :  Genetex
undefined
Anti-FAAP24 Rabbit Polyclonal Antibody
  89322-950
 :  Genetex
 :  GTX117277

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Fanconi anemia core complex 24 kDa subunit
  • Antigen Symbol:
    FAAP24
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    91442
  • Antigen Synonyms:
    C19orf40
  • Storage Buffer:
    0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Storage Temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    0.94 mg/ml
  • Immunogen:
    Recombinant protein encompassing a sequence within the center region of human FAAP24. The exact sequence is proprietary.
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. No.:
    89322-950
  • Supplier no.:
    GTX117277

 

 

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: FAAP24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human