Anti-RGS1 Rabbit Polyclonal Antibody
Catalog # 89322-498
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Regulator of G-protein Signaling 1
- Antigen Symbol:RGS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Bovine,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:5663
- Antigen Synonyms:B-cell activation protein BL34|BL34|IER1|regulator of G-protein signalling 1|1R20|immediate-early response 1|early response protein 1R20|IR20|Regulator of G-protein signaling 1|B-cell specific
- Storage Buffer:0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:0.53 mg/ml
- Immunogen:Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human Presenilin 1. The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89322-498
- Supplier no.:GTX116016
Specifications
About this item
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: RGS1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Bovine, Dog, Rat