To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Chromobox Homolog 5
- Antigen Symbol:CBX5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Dog,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:8854
- Antigen Synonyms:heterochromatin protein 1-alpha|2610029O15Rik|antigen p25|C75991|Hp1a|HP1A|Hp1alpha|HP1Hs alpha|HEL25|HP1 alpha homolog|HP1|Drosophila)|chromobox protein homolog 5|heterochromatin protein 1 homolog alpha|chromobox homolog 5 (HP1 alpha homolog|HP1-ALPHA
- Storage Buffer:0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:0.73 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human ALDH1A2. The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89322-038
- Supplier no.:GTX115147
Specifications
About this item
Suggested starting dilutions are as follows: WB: 1:500-1:20000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: CBX5 / HP1-Alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Dog