Anti-LDHC Mouse Monoclonal Antibody [clone: HBe5]
Catalog # 89336-538
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Lactate dehydrogenase C
- Antigen Symbol:LDHC
- Clonality:Monoclonal
- Clone:HBe5
- ELISA:Yes
- Host:Mouse
- Reactivity:Human,Mouse
- Size:100 µg
- Epitope:221-233 aa
- Antigen Synonyms:CT32|LDHX|LDH3
- Cat. No.:89336-538
- Supplier no.:GTX11445
Specifications
About this item
Type: Primary
Antigen: LDHC
Clonality: Monoclonal
Clone: HBe5
Conjugation:
Epitope: 221-233 aa
Host: Mouse
Isotype:
Reactivity: Human, Mouse