Order Entry
United States
ContactUsLinkComponent
 
Anti-OR51Q1 Rabbit Polyclonal Antibody
  89320-414
 :  Genetex
undefined
Anti-OR51Q1 Rabbit Polyclonal Antibody
  89320-414
 :  Genetex
 :  GTX109792
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Olfactory Receptor, Family 51, Subfamily Q Member 1
  • Antigen Symbol:
    OR51Q1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    1009
  • Antigen Synonyms:
    OB|OSF-4|CDHOB|CAD11
  • Storage Buffer:
    0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Storage Temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    0.8mg/ml
  • Immunogen:
    Recombinant protein encompassing a sequence within the center region of human CDH11. The exact sequence is proprietary.
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. No.:
    89320-414
  • Supplier no.:
    GTX109792

 

 

Rabbit Polyclonal antibody to OB-cadherin (cadherin 11, type 2, OB-cadherin (osteoblast))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: OR51Q1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human