Anti-DYNC1LI1 Rabbit Polyclonal Antibody
Catalog # 89317-208
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Dynein cytoplasmic 1 light intermediate chain 1
- Antigen Symbol:DYNC1LI1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 μg
- Gene ID:2021
- Antigen Synonyms:DNCLI1|LIC1
- Storage Buffer:Preservative: 0.02% Sodium Azide; Constituents: PBS
- Storage Temperature:Upon receipt - Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1mg/ml
- Immunogen:Synthetic peptide: GGPRGPGELAKYGLP, corresponding to amino acids 55-70 of Human Endonuclease G.
- Purification:Immunogen affinity purified
- Cat. No.:89317-208
- Supplier no.:GTX10522
Specifications
About this item
Rabbit Polyclonal antibody to EndoG
WB: Use at a concentration of 1 - 2 ?g/ml. IHC-P: Use at a concentration of 10-
15?g/ml. Detects a band of approximately 35 kDa (predicted molecular weight: 35 kDa), determined using human HepG2 (hepatoblastoma) cell lysate. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: DYNC1LI1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human