To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Cytokeratin 8
- Antigen Symbol:CK8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 μg
- Environmentally Preferable:
- Neutralisation:Yes
- Gene ID:2252
- Antigen Synonyms:CYK8|Keratin|CK-8|KRT8|type II cytoskeletal 8|Keratin-8|K8|Type-II keratin Kb8|Cytokeratin-8
- Storage Buffer:Preservative: NoneConstituents: PBS
- Concentration:0.25mg/ml
- Immunogen:Recombinant full length protein (Human).
- Purification:Immunogen affinity purified
- Cat. No.:89315-996
- Supplier no.:GTX10423
Specifications
About this item
Goat polyclonal to KGF
ELISA: Use at a concentration of 0.5 - 1.0 ?g/ml. Neut: Use at a concentration of 0.1 - 100 ?g/ml. WB: Use at a concentration of 0.1 - 0.2 ?g/ml. Predicted molecular weight: 22.5 kDa. The Neutralization Dose 50 (ND50) for anti-human FGF7 (KGF) is approximately 6 to 12 ?g/ml in the presence of 125 ng/ml of recombinant human FGF7 using the human 4MBr-5 cell line. The ND50 is the concentration of antibody required to yield one-half maximal inhibition of the cytokine activity on a responsive cell line, when the cytokine is present at a concentration just high enough to elicit a maximum response. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: CK8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: