Anti-KIR2DL5B Rabbit Polyclonal Antibody
89397-336
: Abgent
- Antibody Type:Primary
- Antigen Name:Killer cell immunoglobulin-like Receptor two Domains long cytoplasmic tail 5B
- Antigen Symbol:KIR2DL5B
- Clonality:Polyclonal
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Antigen Synonyms:KIR2DL5|KIR2DLX|KIR2DL5.2
- Cat. No.:89397-336
- Supplier no.:AP8909C
Type: Primary
Antigen: KIR2DL5B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human