Anti-JUN Rabbit Polyclonal Antibody
100 µg
Catalog # 89387-018
Supplier: Abgent
Supplier #: AP3072A
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Type: Primary
Antigen: JUN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Documents
AP3072a.pdf
Datasheet
Specifications
- Size:100 µg
- Cat. No.:89387-018
- Clonality:Polyclonal
- Reactivity:Human
- Western Blot:Yes
- Antibody Type:Primary
- Antigen Symbol:JUN
- Host:Rabbit
- ImmunoChemistry:Yes
- Antigen Synonyms:Transcription factor AP-1|V-jun avian sarcoma virus 17 oncogene homolog|JUN|AP1|p39|Proto-oncogene c-jun|Activator protein 1
- Antigen Name:c-Jun
- ELISA:Yes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



