Anti-SIGLEC12 Rabbit Polyclonal Antibody
89384-726
: Abgent
- Antibody Type:Primary
- Antigen Name:Sialic Acid Binding Ig-like Lectin 12
- Antigen Symbol:SIGLEC12
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Antigen Synonyms:mSiglec-E|Siglec9|SIGLECL1|Siglec5|Siglec12|Cd170|Siglecl1
- Cat. No.:89384-726
- Supplier no.:AP1634B
Type: Primary
Antigen: SIGLEC12
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse