Order Entry
United States
ContactUsLinkComponent
 
Anti-MMP28 Rabbit Polyclonal Antibody
  89383-334
 :  Abgent
Anti-MMP28 Rabbit Polyclonal Antibody
  89383-334
 :  Abgent
 :  AP13734C

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Matrix Metalloproteinase 28
  • Antigen Symbol:
    MMP28
  • Clonality:
    Polyclonal
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Antigen Synonyms:
    MM28|MMP25|MMP-28|EPILYSIN|MMP-25
  • Cat. No.:
    89383-334
  • Supplier no.:
    AP13734C

 

 

Type: Primary
Antigen: MMP28
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human