Anti-DNMT3A Rabbit Polyclonal Antibody
Catalog # 89376-600
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:DNA (cytosine-5-)-methyltransferase 3 alpha
- Antigen Symbol:DNMT3A
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Antigen Synonyms:2.1.1.37|DNA (cytosine-5)-methyltransferase 3A|DNA MTase HsaIIIA|M.HsaIIIA|Dnmt3a|DNA methyltransferase HsaIIIA
- Cat. No.:89376-600
- Supplier no.:AP1023B
Specifications
About this item
Type: Primary
Antigen: DNMT3A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human