Anti-UBTD1 Mouse Monoclonal Antibody [clone: 189CT14.5.4]
Catalog # 89375-818
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Ubiquitin Domain Containing 1
- Antigen Symbol:UBTD1
- Clonality:Monoclonal
- Clone:189CT14.5.4
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:100 µL
- Antigen Synonyms:UBTD 1|Ubiquitin domain containing 1|UBTD1_HUMAN.|UBTD1|Ubiquitin domain-containing protein 1|Ubiquitin domain containing protein 1
- Cat. No.:89375-818
- Supplier no.:AM1873A
Specifications
About this item
Type: Primary
Antigen: UBTD1
Clonality: Monoclonal
Clone: 189CT14.5.4
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human