ContactUsLinkComponent
Three-Way Stopcock Luer Valve, ALA Scientific Instruments
Three-Way Stopcock Luer Valve, ALA Scientific Instruments
Catalog # 100133-888
Three-Way Stopcock Luer Valve, ALA Scientific Instruments
Catalog # 100133-888
Supplier Number:  3WLV
Specifications
  • Description:
    Manual 3-Way Luer Valve
  • Environmentally Preferable:
  • Cat. No.:
    100133-888
  • Supplier no.:
    3WLV
Specifications
About this item
Manual luer valves allow for reservoir solution output control and provide an easy way to input solution into a reservoir.