Order Entry
United States
Orders LinkContactUsLinkComponent
 

Mercury (II) sulfide

Mercury (II) sulfide
Supplier:  
Certificates
Mercury (II) sulfide

0 Options Available Below

Product Details & Documents

CAS: 1344-48-5; EC No: 215-696-3; MDL No: MFCD00011046; RTECS: OX0720000 Powder; Molecular Formula: HgS; MW: 232.66 Melting Point: 584° (sublimes) Density (g/mL): 7.7

  • RTECS: OX0720000
  • Powder
  • CAS Number:1344-48-5
  • Formula:HgS
  • MDL Number:MFCD00011046
  • Molecular Weight:232.66 g/mol
  • Melting Pt:584 °C (subl.)
  • Density:7.7 g/mL
  • EINECS:215-696-3
  • UN:2025
  • ADR:6.1,II
Product Family Options