$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United States
ContactUsLinkComponent
Standards
Environmental Analysis Standards
Hydrazine, Standard Solution, N₂H₄ 100 µg/ml
Hydrazine, Standard Solution, N₂H₄ 100 µg/ml
Hydrazine, Standard Solution, N₂H₄ 100 µg/ml
Catalog # BT144320-500ML
Supplier:
BeanTown Chemical
CAS Number:
SDS
Expand image
Hydrazine, Standard Solution, N₂H₄ 100 µg/ml
Catalog # BT144320-500ML
Supplier:
BeanTown Chemical
Supplier Number:
144320-500ML
CAS Number:
SDS
Specifications
Pack type:
HDPE Plastic bottle
Size:
500 ml
MDL No.:
MFCD00011417
Cat. No.:
BT144320-500ML
Supplier no.:
144320-500ML
Specifications
About this item
Product Details
Documentation
Product Details
Documentation