Acetate, Ion Chromatography Standard Solution, CH₃CO₂⁻ 1,000 µg/ml
Supplier: BeanTown Chemical
Certificates
About this item
Specifications
- MDL No.:MFCD00282007
- Solubility:Fully miscible
- Physical Form:Liquid
Specifications
Frequently Bought Together