ContactUsLinkComponent
Nafion® ≥0.92 meq/g ion exchange capacity, Thickness 0.05 mm
Nafion® ≥0.92 meq/g ion exchange capacity, Thickness 0.05 mm
Supplier:  Avantor
Certificates
Nafion® ≥0.92 meq/g ion exchange capacity, Thickness 0.05 mm
Supplier:  Avantor