Order Entry
United States
Orders LinkContactUsLinkComponent
DL-α-Tocopherol (Vitamin E) ≥97%

DL-α-Tocopherol (Vitamin E) ≥97%

DL-α-Tocopherol (Vitamin E) ≥97%
Supplier:  BeanTown Chemical
Certificates

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

CAS: 10191-41-0; EC No: 233-466-0; MDL No: MFCD00072051; RTECS: GA8746000 Liquid; Molecular Formula: C29H50O2; MW: 430.72 Flash point: 110°C (230°F) Density (g/mL): 0.95 Light Sensitive

  • RTECS: GA8746000
  • Light Sensitive
  • Liquid
Synonyms:  (±)-α-Tocopherol, Vitamin E Alcohol, all-rac-α-Tocopherol, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-ol, DL-alpha-Tocopherol, Vitamin E
  • CAS Number:10191-41-0
  • Formula:C₂₉H₅₀O₂
  • MDL Number:MFCD00072051
  • Molecular Weight:430.71 g/mol
  • Density:0.95 g/mL
  • Flash Pt:>110 °C (>230 °F)
  • EINECS:233-466-0

Documents

Specifications for Product Family

Specification Test Results

  • Assay:
    97%
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 50 g
    Log in for availability
    Log in for availability
    Each (50g)
    Each (50g) - $44.80
    $44.80
    Each (50g)
       $44.80

    Supplier Number: 136635-50G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 250 g
    Log in for availability
    Log in for availability
    Each (250g)
    Each (250g) - $111.00
    $111.00
    Each (250g)
       $111.00

    Supplier Number: 136635-250G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 1 kg
    Log in for availability
    Log in for availability
    Each (1kg)
    Each (1kg) - $352.00
    $352.00
    Each (1kg)
       $352.00

    Supplier Number: 136635-1KG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.