Order Entry
United States
ContactUsLinkComponent
Dimethyl sulfoxide-[D₆] ≥100% (Isotopic) contains 0.03% TMS
Dimethyl sulfoxide-[D₆] ≥100% (Isotopic) contains 0.03% TMS
Supplier:  BeanTown Chemical
Certificates
Dimethyl sulfoxide-[D₆] ≥100% (Isotopic) contains 0.03% TMS
Supplier:  BeanTown Chemical

Specification Test Results

  • Assay:
    100%

Specification Test Results

Related Information

Frequently Bought Together