Fluoride, Ion chromatography standard solution, F⁻ 1000 µg/ml
Supplier: BeanTown Chemical
Certificates
About this item
Specifications
- MDL No.:MFCD00144315
- Solubility:Fully miscible
- Physical Form:Liquid
Specifications
Frequently Bought Together