Potassium permanganate 0.1 N in water volumetric solution
Supplier: BeanTown Chemical
Certificates
About this item
CAS Number:7722-64-7
:
Formula:KMnO₄
MDL Number:MFCD00011364
:158.03 g/mol
Boiling Pt:100 °C
Melting Pt:0 °C
Density:1 g/mL
EINECS:231-760-3
UN:3082
ADR:9,III
Frequently Bought Together