Order Entry
United States
ContactUsLinkComponent
 
Tungsten, Plasma Standard Solution, W 1000 µg/ml
 
Tungsten, Plasma Standard Solution, W 1000 µg/ml

 

  • MDL No.:
    MFCD00011461
  • Physical Form:
    Liquid

 

 

Frequently Bought Together